Imprimer

Anti-VEGFA Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17696-50EA 572 EUR
ABCAAB17696-50
Anti-VEGFA Goat polyclonal antibody
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR