Imprimer

Anti-BCAR1 Mouse monoclonal antibody [clone: SPM302]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17745-7EA 579 EUR
ABCAAB17745-7
Anti-BCAR1 Mouse monoclonal antibody [clone: SPM302]
Type: Primary
Antigen: BCAR1 (breast cancer anti-estrogen resistance 1)
Clonality: monoclonal
Clone: SPM302
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR