Imprimer

Anti-Prostatic Acid Phosphatase Mouse monoclonal antibody [clone: SPM312]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17793-7EA 579 EUR
ABCAAB17793-7
Anti-Prostatic Acid Phosphatase Mouse monoclonal antibody [clone: SPM312]
Type: Primary
Antigen: Prostatic Acid Phosphatase
Clonality: monoclonal
Clone: SPM312
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR