Imprimer

Anti-HSPA13 Mouse monoclonal antibody [clone: SPM254]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17850-7EA 0 EUR
ABCAAB17850-7
Anti-HSPA13 Mouse monoclonal antibody [clone: SPM254]
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: SPM254
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR