Imprimer

Anti-MMP7 Mouse monoclonal antibody [clone: SPM294]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17854-7EA 579 EUR
ABCAAB17854-7
Anti-MMP7 Mouse monoclonal antibody [clone: SPM294]
Type: Primary
Antigen: MMP7 (matrix metallopeptidase 7 (matrilysin, uterine))
Clonality: monoclonal
Clone: SPM294
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR