Imprimer

Anti-Cdc7 Kinase Mouse monoclonal antibody [clone: SPM171]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17880-250EA 593 EUR
ABCAAB17880-250
Anti-Cdc7 Kinase Mouse monoclonal antibody [clone: SPM171]
Type: Primary
Antigen: Cdc7 Kinase
Clonality: monoclonal
Clone: SPM171
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR