Imprimer

Anti-BRWD1 Chicken polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17907-250EA 565 EUR
ABCAAB17907-250
Anti-BRWD1 Chicken polyclonal antibody
Type: Primary
Antigen: BRWD1 (bromodomain and WD repeat domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR