Imprimer

Anti-C5 Mouse monoclonal antibody unconjugated [clone: 11F6]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17930-100EA 862 EUR
ABCAAB17930-100
Anti-C5 Mouse monoclonal antibody unconjugated [clone: 11F6]
Anticorps
Type: Primary
Antigen: C5 (complement component 5)
Clonality: monoclonal
Clone: 11F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR