Imprimer

Anti-Factor B Mouse Monoclonal Antibody [clone: 9B8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17932-100EA 705 EUR
ABCAAB17932-100
Anti-Factor B Mouse Monoclonal Antibody [clone: 9B8]
Anticorps
Type: Primary
Antigen: Factor B
Clonality: monoclonal
Clone: 9B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR