Imprimer

Anti-Surfactant Protein B (Mature) Mouse monoclonal antibody [clone: SPM219]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17946EA 579 EUR
ABCAAB17946
Anti-Surfactant Protein B (Mature) Mouse monoclonal antibody [clone: SPM219]
Type: Primary
Antigen: Surfactant Protein B (Mature)
Clonality: monoclonal
Clone: SPM219
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR