Imprimer

Anti-HLTF Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17984-50EA 729 EUR
ABCAAB17984-50
Anti-HLTF Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: HLTF (helicase-like transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR