Imprimer

Anti-FEN1 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17993-100EA 684 EUR
ABCAAB17993-100
Anti-FEN1 Rabbit polyclonal antibody
Type: Primary
Antigen: FEN1 (flap structure-specific endonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR