Imprimer

Anti-Frequenin Chicken polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18060-50EA 550 EUR
ABCAAB18060-50
Anti-Frequenin Chicken polyclonal antibody
Type: Primary
Antigen: Frequenin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR