Imprimer

Anti-FLG Mouse monoclonal antibody [clone: SPM181]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18098-7EA 579 EUR
ABCAAB18098-7
Anti-FLG Mouse monoclonal antibody [clone: SPM181]
Type: Primary
Antigen: FLG (filaggrin)
Clonality: monoclonal
Clone: SPM181
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR