Imprimer

Anti-calf intestinal Alkaline Phosphatase Mouse Monoclonal Antibody [clone: AP1B9]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18110-50EA 691 EUR
ABCAAB18110-50
Anti-calf intestinal Alkaline Phosphatase Mouse Monoclonal Antibody [clone: AP1B9]
Anticorps
Type: Primary
Antigen: calf intestinal Alkaline Phosphatase
Clonality: monoclonal
Clone: AP1B9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR