Imprimer

Anti-HIV2 p24 Mouse monoclonal antibody [clone: 8.F.156]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18145-25EA 550 EUR
ABCAAB18145-25
Anti-HIV2 p24 Mouse monoclonal antibody [clone: 8.F.156]
Type: Primary
Antigen: HIV2 p24
Clonality: monoclonal
Clone: 8.F.156
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR