Imprimer

Anti-HA tag Mouse Monoclonal Antibody [clone: HA.C5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18181-100EA 775 EUR
ABCAAB18181-100
Anti-HA tag Mouse Monoclonal Antibody [clone: HA.C5]
Anticorps
Type: Primary
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: HA.C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR