Imprimer

Anti-H3F3AP6 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18209-100EA 613 EUR
ABCAAB18209-100
Anti-H3F3AP6 Rabbit polyclonal antibody
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR