Imprimer

Anti-CD63 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-259]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18236-50EA 347 EUR
ABCAAB18236-50
Anti-CD63 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-259]
Anticorps
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone: MEM-259
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR