Imprimer

Anti-CCNB2 Mouse monoclonal antibody unconjugated [clone: X29.2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18250-100EA 751 EUR
ABCAAB18250-100
Anti-CCNB2 Mouse monoclonal antibody unconjugated [clone: X29.2]
Anticorps
Type: Primary
Antigen: CCNB2 (cyclin B2)
Clonality: monoclonal
Clone: X29.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR