Imprimer

Anti-CD80 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-233]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18279-50EA 345 EUR
ABCAAB18279-50
Anti-CD80 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: MEM-233]
Anticorps
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: monoclonal
Clone: MEM-233
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR