Imprimer

Anti-LCP2 Mouse monoclonal antibody [clone: H76]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18335-100EA 550 EUR
ABCAAB18335-100
Anti-LCP2 Mouse monoclonal antibody [clone: H76]
Type: Primary
Antigen: LCP2 (lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa))
Clonality: monoclonal
Clone: H76
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR