Imprimer

Anti-RELN Mouse monoclonal antibody [clone: G10]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18570-50EA 596 EUR
ABCAAB18570-50
Anti-RELN Mouse monoclonal antibody [clone: G10]
Type: Primary
Antigen: RELN (reelin)
Clonality: monoclonal
Clone: G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR