Imprimer

Anti-RuvB Rabbit polyclonal antibody unconjugated

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18581-100EA 635 EUR
ABCAAB18581-100
Anti-RuvB Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: RuvB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR