Imprimer

Anti-Cytokeratin 6 Mouse Monoclonal Antibody [clone: Ks6.KA12]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18586-500EA 928 EUR
ABCAAB18586-500
Anti-Cytokeratin 6 Mouse Monoclonal Antibody [clone: Ks6.KA12]
Anticorps
Type: Primary
Antigen: Cytokeratin 6
Clonality: monoclonal
Clone: Ks6.KA12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR