Imprimer

Anti-TUBB Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18587-100EA 572 EUR
ABCAAB18587-100
Anti-TUBB Rabbit polyclonal antibody
Type: Primary
Antigen: TUBB (tubulin, beta class I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR