Imprimer

Anti-HSV1 gD Mouse Monoclonal Antibody [clone: 1-I-9]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18638-100EA 698 EUR
ABCAAB18638-100
Anti-HSV1 gD Mouse Monoclonal Antibody [clone: 1-I-9]
Anticorps
Type: Primary
Antigen: HSV1 gD
Clonality: monoclonal
Clone: 1-I-9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR