Imprimer

Anti-IL8 Mouse Monoclonal Antibody [clone: 807]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18672-250EA 733 EUR
ABCAAB18672-250
Anti-IL8 Mouse Monoclonal Antibody [clone: 807]
Anticorps
Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: monoclonal
Clone: 807
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR