Imprimer

Anti-PTPN6 Mouse monoclonal antibody [clone: PTY15]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB18708-100EA 572 EUR
ABCAAB18708-100
Anti-PTPN6 Mouse monoclonal antibody [clone: PTY15]
Type: Primary
Antigen: PTPN6 (protein tyrosine phosphatase, non-receptor type 6)
Clonality: monoclonal
Clone: PTY15
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR