Imprimer

Anti-Kisspeptin Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19028-100EA 729 EUR
ABCAAB19028-100
Anti-Kisspeptin Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Kisspeptin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR