Imprimer

Anti-USH1C Goat Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19045-100EA 658 EUR
ABCAAB19045-100
Anti-USH1C Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: USH1C (Usher syndrome 1C (autosomal recessive, severe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR