Imprimer

Anti-LSM4 Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19101-50EA 766 EUR
ABCAAB19101-50
Anti-LSM4 Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: LSM4 (LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR