Imprimer

Anti-CDC25A Rabbit Polyclonal Antibody (Agarose)

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19204-100EA 698 EUR
ABCAAB19204-100
Anti-CDC25A Rabbit Polyclonal Antibody (Agarose)
Anticorps
Type: Primary
Antigen: CDC25A (cell division cycle 25A)
Clonality: polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR