Imprimer

Anti-CDKL3 Rabbit polyclonal antibody unconjugated

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19251-100EA 798 EUR
ABCAAB19251-100
Anti-CDKL3 Rabbit polyclonal antibody unconjugated
Anticorps
Type: Primary
Antigen: CDKL3 (cyclin-dependent kinase-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR