Imprimer

Anti-VSV-G Tag Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19257-250EA 698 EUR
ABCAAB19257-250
Anti-VSV-G Tag Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: VSV-G Tag (VSV-G tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR