Imprimer

Anti-UL42 Mouse Monoclonal Antibody [clone: 13C9]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19298-50EA 838 EUR
ABCAAB19298-50
Anti-UL42 Mouse Monoclonal Antibody [clone: 13C9]
Anticorps
Type: Primary
Antigen: UL42
Clonality: monoclonal
Clone: 13C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR