Imprimer

Anti-CASR Mouse Monoclonal Antibody [clone: 5C10+ADD]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19347-100EA 757 EUR
ABCAAB19347-100
Anti-CASR Mouse Monoclonal Antibody [clone: 5C10+ADD]
Anticorps
Type: Primary
Antigen: CASR (calcium-sensing receptor)
Clonality: monoclonal
Clone: 5C10+ADD
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR