Imprimer

Anti-HB Goat Polyclonal Antibody (FITC (Fluorescein))

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19361-1EA 698 EUR
ABCAAB19361-1
Anti-HB Goat Polyclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: HB (hemoglobin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR