Imprimer
Anti-HSV tag Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB19392-100EA 878 EUR
ABCAAB19392-100
Anti-HSV tag Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Primary
Antigen: HSV tag
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Commander maintenant