Imprimer

Anti-HCN1 Rabbit polyclonal antibody unconjugated

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19405-100EA 704 EUR
ABCAAB19405-100
Anti-HCN1 Rabbit polyclonal antibody unconjugated
Anticorps
Type: Primary
Antigen: HCN1 (hyperpolarization activated cyclic nucleotide-gated potassium channel 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR