Imprimer

Anti-FAS Mouse monoclonal antibody [clone: 5F-7]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19454-50EA 550 EUR
ABCAAB19454-50
Anti-FAS Mouse monoclonal antibody [clone: 5F-7]
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: monoclonal
Clone: 5F-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR