Imprimer

Anti-FAS Mouse monoclonal antibody [clone: IPO-4]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19456-50EA 540 EUR
ABCAAB19456-50
Anti-FAS Mouse monoclonal antibody [clone: IPO-4]
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: monoclonal
Clone: IPO-4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR