Imprimer

Anti-BCL11A Mouse monoclonal antibody unconjugated [clone: 14B5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19487-100EA 795 EUR
ABCAAB19487-100
Anti-BCL11A Mouse monoclonal antibody unconjugated [clone: 14B5]
Anticorps
Type: Primary
Antigen: BCL11A (B-cell CLL/lymphoma 11A (zinc finger protein))
Clonality: monoclonal
Clone: 14B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR