Imprimer

Anti-CEACAM 5 + 6 Mouse monoclonal antibody [clone: 198]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19563-100EA 0 EUR
ABCAAB19563-100
Anti-CEACAM 5 + 6 Mouse monoclonal antibody [clone: 198]
Type: Primary
Antigen: CEACAM 5 + 6
Clonality: monoclonal
Clone: 198
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR