Imprimer

Anti-BNP Rabbit polyclonal antibody unconjugated

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19645-50EA 720 EUR
ABCAAB19645-50
Anti-BNP Rabbit polyclonal antibody unconjugated
Anticorps
Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR