Imprimer

Anti-CD19 Mouse monoclonal antibody [clone: BC-3]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19650-500EA 572 EUR
ABCAAB19650-500
Anti-CD19 Mouse monoclonal antibody [clone: BC-3]
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: BC-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR