Imprimer

Anti-ITGAM Mouse monoclonal antibody Biotin [clone: 44]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19674-50EA 285 EUR
ABCAAB19674-50
Anti-ITGAM Mouse monoclonal antibody Biotin [clone: 44]
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone: 44
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR