Imprimer

Anti-FABP4 Mouse monoclonal antibody [clone: 5B5]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19724-500EA 572 EUR
ABCAAB19724-500
Anti-FABP4 Mouse monoclonal antibody [clone: 5B5]
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: monoclonal
Clone: 5B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR