Imprimer

Anti-ALP Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19734-200EA 550 EUR
ABCAAB19734-200
Anti-ALP Rabbit polyclonal antibody
Type: Primary
Antigen: ALP (Alkaline Phosphatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR