Imprimer

Anti-ABCC11 Mouse Monoclonal Antibody [clone: 2C5/4]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB19860-50EA 704 EUR
ABCAAB19860-50
Anti-ABCC11 Mouse Monoclonal Antibody [clone: 2C5/4]
Anticorps
Type: Primary
Antigen: ABCC11 (ATP-binding cassette, sub-family C (CFTR/MRP), member 11)
Clonality: monoclonal
Clone: 2C5/4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR